Erbium ≥99.9%, chips

Supplier: STREM CHEMICALS INC MS

93-6831-5G 93-6831-25G
100200-792EA 140.12 USD
100200-792 100200-790
Erbium ≥99.9%, chips
Erbium
Formula: Er
MW: 167.26 g/mol
Boiling Pt: 2863 °C
Melting Pt: 1529 °C
Density: 9,006
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR