Erbium(III) oxide ≥99.9% (Er basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dierbium trioxide, Erbium oxide

93-6810-5G 93-6810-100G 93-6810-25G
100204-416EA 53.78 USD
100204-416 100209-196 100200-788
Erbium(III) oxide ≥99.9% (Er basis)
Erbium(III) oxide
Formula: Er₂O₃
MW: 382.52 g/mol
Melting Pt: 2420 °C
Density: 7.36 g/cm³ (20 °C)
MDL Number: MFCD00010991
CAS Number: 12061-16-4
EINECS: 235-045-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR