Erbium trichloride hydrate ≥99.9% (Er basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6804-100G 93-6804-25G
100199-302EA 311.38 USD
100199-302 100196-814
Erbium trichloride hydrate ≥99.9% (Er basis)
Erbium trichloride hydrate
Formula: Cl₃Er·H₂O
MW: 291.63 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00010989
CAS Number: 19423-85-9
EINECS: 233-385-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR