Erbium trichloride, anhydrous ≥99.9% (Er basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6803-25G 93-6803-5G
100207-894EA 338.28 USD
100207-894 100207-896
Erbium trichloride, anhydrous ≥99.9% (Er basis)
Erbium trichloride
Formula: Cl₃Er
MW: 273.62 g/mol
Boiling Pt: 1500 °C
Melting Pt: 774 °C
Density: 4.1 g/cm³ (20 °C)
MDL Number: MFCD00010989
CAS Number: 10138-41-7
EINECS: 233-385-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR