Holmium ≥99.9%, ingot
Supplier: STREM CHEMICALS INC MS
93-6732-5G
93-6732-25G
100205-736EA
229.29
USD
100205-736
100202-574
Holmium ≥99.9%, ingot
Holmium
Formula:
Ho MW: 164.93 Boiling Pt: 2695 °C Melting Pt: 1474 °C Density: 8.795 |
MDL Number:
MFCD00011049 CAS Number: 7440-60-0 EINECS: 231-169-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...