Holmium ≥99%, chips

Supplier: STREM CHEMICALS INC MS

93-6731-2G 93-6731-10G
100211-286EA 101.91 USD
100211-286 100206-164
Holmium ≥99%, chips
Holmium
Formula: Ho
Boiling Pt: 2695 °C
Melting Pt: 1474 °C
Density: 8,795
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR