Holmium ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-6730-10G 93-6730-2G
100203-564EA 697.78 USD
100203-564 100207-892
Holmium ≥99.9%, powder
Holmium
Formula: Ho
MW: 164.93 g/mol
Boiling Pt: 2695 °C (1013 hPa)
Melting Pt: 1474 °C
Density: 8.7947 g/cm³ (20 °C)
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0
UN: 3089
ADR: 4.1,II
Merck Index: 12,04763

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR