Holmium(III) oxide ≥99.9% (Ho basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Diholmium trioxide, Holmium oxide
93-6713-5G
93-6713-25G
100210-532EA
67.94
USD
100210-532
100207-890
Holmium(III) oxide ≥99.9% (Ho basis)
Holmium(III) oxide
Formula:
Ho₂O₃ MW: 377.86 g/mol Boiling Pt: 2600 Melting Pt: 2400 Density: 8.36 |
MDL Number:
MFCD00011053 CAS Number: 12055-62-8 EINECS: 235-015-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...