Dysprosium ≥99.9%, foil
Supplier: STREM CHEMICALS INC MS
93-6638-50X50MM
93-6638-25X25MM
100207-488EA
653.91
USD
100207-488
100198-438
Dysprosium ≥99.9%, foil
Dysprosium
Formula:
Dy MW: 162.5 Boiling Pt: 2335 °C Melting Pt: 1409 °C Density: 8.54 |
MDL Number:
MFCD00010982 CAS Number: 7429-91-6 EINECS: 231-073-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...