Dysprosium ≥99.9%, chips

Supplier: STREM CHEMICALS INC MS

93-6636-5G 93-6636-25G
100211-284EA 140.12 USD
100211-284 100202-260
Dysprosium ≥99.9%, chips
Dysprosium
Formula: Dy
MW: 162.5
Boiling Pt: 2335 °C
Melting Pt: 1409 °C
Density: 8.54
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR