Dysprosium ≥99.9%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-6635-25G
93-6635-5G
100210-892EA
443.02
USD
100210-892
100210-528
Dysprosium ≥99.9%, powder
Dysprosium
Formula:
Dy MW: 162.5 Boiling Pt: 2335 °C Melting Pt: 1409 °C Density: 8.54 |
MDL Number:
MFCD00010982 CAS Number: 7429-91-6 EINECS: 231-073-9 UN: 3089 ADR: 4.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...