Dysprosium trichloride hexahydrate ≥99.9% (Dy basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Dysprosium (III) chloride hexahydrate, Dysprosium(III) chloride hydrate
93-6628-25G
93-6628-100G
100204-410EA
89.17
USD
100204-410
100202-258
Dysprosium trichloride hexahydrate ≥99.9% (Dy basis)
Dysprosium trichloride hexahydrate
Formula:
Cl₃Dy·6H₂O MW: 376.95 g/mol Melting Pt: 163 °C |
MDL Number:
MFCD00010984 CAS Number: 15059-52-6 EINECS: 233-039-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...