Dysprosium trichloride, anhydrous ≥99.9% (dye basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6607-2G 93-6607-50G 93-6607-10G
100199-162EA 43.88 USD
100199-162 100207-304 100208-222
Dysprosium trichloride, anhydrous ≥99.9% (dye basis)
Dysprosium trichloride
Formula: Cl₃Dy
MW: 268.86 g/mol
Melting Pt: 642 - 652 °C
Density: 3.67 g/cm³ (0 °C)
MDL Number: MFCD00010984
CAS Number: 10025-74-8
EINECS: 233-039-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR