Terbium ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-6515-5G 93-6515-1G
100197-032EA 452.92 USD
100197-032 100208-588
Terbium ≥99.9%, powder
Terbium
Formula: Tb
Boiling Pt: 3041 °C
Melting Pt: 1360 °C
Density: 8.234
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR