Terbium trinitrate hexahydrate ≥99.9% (Tb basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-6508-5G
93-6508-25G
100209-186EA
77.85
USD
100209-186
100207-138
Terbium trinitrate hexahydrate ≥99.9% (Tb basis)
Terbium trinitrate hexahydrate
Formula:
N₃O₉Tb·6H₂O Melting Pt: 89,3 °C |
MDL Number:
MFCD00149868 CAS Number: 13451-19-9 EINECS: 233-138-7 UN: 1477 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...