Gadolinium trifluoride, anhydrous ≥99.9% (Gd basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Gadolinium (III) fluoride, Gadolinium(III) bromide

93-6422-50G 93-6422-10G
100202-824EA 315.63 USD
100202-824 100205-726
Gadolinium trifluoride, anhydrous ≥99.9% (Gd basis)
Gadolinium trifluoride
Formula: F₃Gd
MW: 214.25 g/mol
Melting Pt: 1231 °C
MDL Number: MFCD00016113
CAS Number: 13765-26-9
EINECS: 237-369-4
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR