Gadolinium trinitrate hexahydrate ≥99.9% (Gd basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Gadolinium (III) nitrate hexahydrate

93-6417-25G 93-6417-100G
100196-800EA 72.18 USD
100196-800 100209-176
Gadolinium trinitrate hexahydrate ≥99.9% (Gd basis)
Gadolinium trinitrate hexahydrate
Formula: GdN₃O₉·6H₂O
MW: 451.36
Melting Pt: 91.5…92 °C
Density: 2.4 g/cm³ (20 °C)
MDL Number: MFCD00149728
CAS Number: 19598-90-4
EINECS: 233-437-2
UN: 1477
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR