Europium(III) fluoride, anhydrous ≥99.99% (Eu basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-6322-1G 93-6322-5G
100205-716EA 69.35 USD
100205-716 100203-548
Europium(III) fluoride, anhydrous ≥99.99% (Eu basis), PURATREM
Europium(III) fluoride
Formula: EuF₃
MW: 208.96 g/mol
Boiling Pt: 2280
Melting Pt: 1390 °C
MDL Number: MFCD00010998
CAS Number: 13765-25-8
EINECS: 237-368-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR