Europium(III) oxide ≥99.99% (Eu basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-6312-5G 93-6312-25G 93-6312-100G
100200-184EA 188.25 USD
100200-184 100209-282 100209-490
Europium(III) oxide ≥99.99% (Eu basis), PURATREM
Europium(III) oxide
Formula: Eu₂O₃
MW: 351.92 g/mol
Melting Pt: 2325
Density: 7.42
MDL Number: MFCD00010997
CAS Number: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,03904

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR