Europium(III) chloride hexahydrate ≥99.9% (Eu basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Europium(III) chloride hydrate, Europium trichloride hexahydrate

93-6306-25G 93-6306-5G
100207-126EA 510.95 USD
100207-126 100207-128
Europium(III) chloride hexahydrate ≥99.9% (Eu basis)
Europium(III) chloride hexahydrate
Formula: Cl₃Eu·6H₂O
MW: 366.41
Melting Pt: 158
Density: 4.89 g/cm³ (20 °C)
MDL Number: MFCD00149701
CAS Number: 13759-92-7
EINECS: 233-040-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR