Europium(III) chloride, anhydrous ≥99.9% (Eu basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6305-2G 93-6305-10G
100202-712EA 116.06 USD
100202-712 100202-710
Europium(III) chloride, anhydrous ≥99.9% (Eu basis)
Europium(III) chloride
Formula: Cl₃Eu
Melting Pt: 850 °C
Density: 4.89 g/cm³ (20 °C)
MDL Number: MFCD00010994
CAS Number: 10025-76-0
EINECS: 233-040-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR