Samarium trichloride hexahydrate ≥99.9% (Sm basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Samarium(III) chloride hexahydrate, Samarium trichloride hexahydrate

93-6225-25G 93-6225-100G
100209-172EA 86.34 USD
100209-172 100210-702
Samarium trichloride hexahydrate ≥99.9% (Sm basis)
Samarium trichloride hexahydrate
Formula: Cl₃Sm·6H₂O
MW: 364.81 g/mol
Density: 2.383 g/cm³ (25 °C)
MDL Number: MFCD00011235
CAS Number: 13465-55-9
EINECS: 233-797-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR