Samarium trinitrate hexahydrate ≥99.9% (Sm basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Samarium (III) nitrate hexahydrate, Samarium(III) nitrate hydrate

93-6207-100G 93-6207-25G
100204-744EA 233.54 USD
100204-744 100209-170
Samarium trinitrate hexahydrate ≥99.9% (Sm basis)
Samarium trinitrate hexahydrate
Formula: N₃O₉Sm·6H₂O
MW: 444.47 g/mol
Melting Pt: 78
Density: 2.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149857
CAS Number: 13759-83-6
EINECS: 233-798-6
UN: 1477
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR