Samarium(III) fluoride ≥99.9% (Sm basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-6205-25G
93-6205-5G
100203-540EA
270.34
USD
100203-540
100205-708
Samarium(III) fluoride ≥99.9% (Sm basis)
Samarium(III) fluoride
Formula:
F₃Sm MW: 207.36 g/mol Boiling Pt: 2323 °C Melting Pt: 1306 °C Density: 4.362 g/cm³ (2327 °C) |
MDL Number:
MFCD00049540 CAS Number: 13765-24-7 EINECS: 237-367-3 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...