Neodymium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS
Warning

93-6045-50X50MM 93-6045-25X25MM
100202-242EA 1582.4 USD
100202-242 100197-922
Neodymium ≥99.9%, foil
Neodymium
Formula: Nd
MW: 144.24
Boiling Pt: 3027 °C
Melting Pt: 1024 °C
Density: 7.00
MDL Number: MFCD00011130
CAS Number: 7440-00-8
EINECS: 231-109-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR