Neodymium(III) oxide ≥99.9% (Nd basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

93-6031-250G 93-6031-50G
100208-730EA 469.91 USD
100208-730 100211-282
Neodymium(III) oxide ≥99.9% (Nd basis)
Neodymium(III) oxide
Formula: Nd₂O₃
MW: 336.48 g/mol
Boiling Pt: 3760 °C (1013 hPa)
Melting Pt: 2272 °C
Density: 7.24 g/cm³ (20 °C)
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,06450

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR