Neodymium trichloride hexahydrate ≥99.9% (Nd basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Neodymium (III) chloride hexahydrate
93-6030-50G
93-6030-10G
93-6030-250G
100200-780EA
101.91
USD
100200-780
100204-394
100199-150
Neodymium trichloride hexahydrate ≥99.9% (Nd basis)
Neodymium trichloride hexahydrate
Formula:
Cl₃Nd·6H₂O MW: 358.69 g/mol Melting Pt: 124 °C Density: 2.28 g/cm³ (20 °C) |
MDL Number:
MFCD00149801 CAS Number: 13477-89-9 EINECS: 233-031-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...