Neodymium trinitrate hexahydrate ≥99.999% (Nd basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Neodymium (III) nitrate hexahydrate
93-6024-5G
93-6024-25G
100197-920EA
66.52
USD
100197-920
100196-790
Neodymium trinitrate hexahydrate ≥99.999% (Nd basis), PURATREM
Neodymium trinitrate hexahydrate
Formula:
N₃NdO₉·6H₂O Melting Pt: ∼74 °C Density: 2.265 g/cm³ (20 °C) |
MDL Number:
MFCD00149799 CAS Number: 16454-60-7 EINECS: 233-153-9 UN: 1477 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...