Neodymium trifluoride, anhydrous ≥99.9% (Nd basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6023-100G 93-6023-25G
100202-818EA 324.12 USD
100202-818 100207-122
Neodymium trifluoride, anhydrous ≥99.9% (Nd basis)
Neodymium trifluoride
Formula: F₃Nd
MW: 201.24
Boiling Pt: 2327
Melting Pt: 1410
Density: 6.506
MDL Number: MFCD00011135
CAS Number: 13709-42-7
EINECS: 237-253-3
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR