Neodymium trichloride, anhydrous ≥99.9% (Nd basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Neodymium (III) chloride

93-6022-10G 93-6022-50G
100201-132EA 138.71 USD
100201-132 100201-134
Neodymium trichloride, anhydrous ≥99.9% (Nd basis)
Neodymium trichloride
Formula: Cl₃Nd
MW: 250.6 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 784 °C
Density: 4.13 g/cm³ (20 °C)
MDL Number: MFCD00011132
CAS Number: 10024-93-8
EINECS: 233-031-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR