Neodymium(III) oxide ≥99.99% (Nd basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

93-6009-25G 93-6009-5G
100205-704EA 359.51 USD
100205-704 100208-586
Neodymium(III) oxide ≥99.99% (Nd basis), PURATREM
Neodymium(III) oxide
Formula: Nd₂O₃
MW: 336.48 g/mol
Boiling Pt: 3760 °C (1013 hPa)
Melting Pt: 2272 °C
Density: 7.24 g/cm³ (20 °C)
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,06450

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR