Dipraseodymium trisulphate hydrate ≥99.9% (Pr basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Praseodymium(III) sulphate hydrate
93-5914-10G
93-5914-50G
100205-700EA
104.74
USD
100205-700
100202-816
Dipraseodymium trisulphate hydrate ≥99.9% (Pr basis)
Dipraseodymium trisulfate hydrate
Formula:
O₁₂Pr₂S₃·xH₂O MW: 570.00 g/mol (anhy) |
MDL Number:
MFCD00149832 CAS Number: 13510-41-3 EINECS: 233-622-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...