Praseodymium(III,IV) oxide ≥99.9% (Pr basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5911-250G
93-5911-50G
93-5911-10G
100200-174EA
437.35
USD
100200-174
100202-232
100207-880
Praseodymium(III,IV) oxide ≥99.9% (Pr basis)
Praseodymium(III,IV) oxide
Formula:
O₁₁Pr₆ MW: 1021.44 g/mol Melting Pt: 2012…2072 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011178 CAS Number: 12037-29-5 EINECS: 234-857-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...