Praseodymium(III,IV) oxide ≥99.99% (Pr basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-5910-25G 93-5910-5G 93-5910-1G
100203-534EA 612.86 USD
100203-534 100199-642 100205-696
Praseodymium(III,IV) oxide ≥99.99% (Pr basis), PURATREM
Praseodymium(III,IV) oxide
Formula: O₁₁Pr₆
MW: 1021.44 g/mol
Melting Pt: 2012…2072 °C
Storage Temperature: Ambient
MDL Number: MFCD00011178
CAS Number: 12037-29-5
EINECS: 234-857-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR