Praseodymium trifluoride ≥99.9% (Pr basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-5906-50G 93-5906-10G
100202-812EA 318.46 USD
100202-812 100205-694
Praseodymium trifluoride ≥99.9% (Pr basis)
Praseodymium trifluoride
Formula: F₃Pr
MW: 197.9 g/mol
Melting Pt: 1395 °C
Density: 6.3 g/cm³ (20 °C)
MDL Number: MFCD00016287
CAS Number: 13709-46-1
EINECS: 237-254-9
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR