Cerium tribromide, anhydrous ≥99.9% (Ce basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cerous bromide, Cerium(III) bromide hydrate, Cerium (III) bromide

93-5861-25G 93-5861-5G
100206-158EA 512.37 USD
100206-158 100204-740
Cerium tribromide, anhydrous ≥99.9% (Ce basis)
Cerium tribromide
Formula: CeBr₃
MW: 379.83 g/mol
Melting Pt: 732 °C
MDL Number: MFCD00016004
CAS Number: 14457-87-5
EINECS: 238-447-0
Merck Index: 12,02039

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR