Cerium trifluoride ≥99.9% (Ce basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Cerous fluoride, Cerium (III) fluoride
93-5825-25G
93-5825-100G
100204-386EA
56.62
USD
100204-386
100208-202
Cerium trifluoride ≥99.9% (Ce basis)
Cerium trifluoride
Formula:
CeF₃ MW: 197.11 Boiling Pt: 1730 Melting Pt: 817 Density: 6.16 |
MDL Number:
MFCD00010932 CAS Number: 7758-88-5 EINECS: 231-841-3 Merck Index: 12,02042 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...