Cerium triacetate hydrate ≥99.9% (Ce basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Acetic acid cerium (III) salt hydrate, Cerium (III) acetate hydrate
93-5801-250G
93-5801-50G
100207-116EA
266.09
USD
100207-116
100205-678
Cerium triacetate hydrate ≥99.9% (Ce basis)
Cerium triacetate hydrate
Formula:
C₆H₉CeO₆·H₂O MW: 335.266 g/mol |
MDL Number:
MFCD00150533 CAS Number: 206996-60-3 EINECS: 208-654-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...