Lanthanum(III) perchlorate hexahydrate

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Lanthanum perchlorate hexahydrate

93-5733-50G 93-5733-10G 93-5733-250G
100197-910EA 130.22 USD
100197-910 100202-216 100200-170
Lanthanum(III) perchlorate hexahydrate
Lanthanum(III) perchlorate hexahydrate
Formula: Cl₃LaO₁₂·6H₂O
MW: 545.35 g/mol
MDL Number: MFCD00016159
CAS Number: 36907-37-6
EINECS: 237-835-7
UN: 1481
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR