Lanthanum(III) nitrate hexahydrate ≥99.99% (La basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

93-5732-25G 93-5732-500G 93-5732-100G
100200-590EA 101.91 USD
100200-590 100202-708 100196-774
Lanthanum(III) nitrate hexahydrate ≥99.99% (La basis), PURATREM
Lanthanum(III) nitrate hexahydrate
Formula: La(NO₃)₃·6H₂O
MW: 433.01 g/mol
Boiling Pt: 126 °C (dec.)
Melting Pt: 40 °C
Density: 2.347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149751
CAS Number: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1,II
Merck Index: 14,05363

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR