Lanthanum(III) oxide ≥99.99% (La basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5716-500G
93-5716-100G
100210-490EA
404.8
USD
100210-490
100205-672
Lanthanum(III) oxide ≥99.99% (La basis), PURATREM
Lanthanum(III) oxide
Formula:
La₂O₃ MW: 325.81 g/mol Boiling Pt: 4200 Melting Pt: 2315 Density: 6.51 Storage Temperature: Ambient |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 EINECS: 215-200-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...