Lanthanum(III) iodide, anhydrous ≥99.9% (La basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-5712-5G 93-5712-1G
100203-526EA 915.75 USD
100203-526 100203-524
Lanthanum(III) iodide, anhydrous ≥99.9% (La basis)
Formula: I₃La
Melting Pt: 772 °C
Density: 5.63
MDL Number: MFCD00049470
CAS Number: 13813-22-4
EINECS: 237-465-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR