Barium hydride ≥99.5% (Ba basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-5654-2G 93-5654-10G
100211-280EA 335.45 USD
100211-280 100211-278
Barium hydride ≥99.5% (Ba basis)
Barium hydride
Formula: H₂Ba
MW: 139.35
Melting Pt: 1200 (dec.)
Density: 4.21
MDL Number: MFCD00003438
CAS Number: 13477-09-3
EINECS: 236-763-3
UN: 3134
ADR: 4.3 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR