Barium stearate, Technical Grade

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Stearic acid barium salt

93-5644-250G 93-5644-1000G
100205-664EA 42.46 USD
100205-664 100210-486
Barium stearate, Technical Grade
Barium stearate
Formula: C₃₆H₇₀BaO₄
MW: 704.28 g/mol
Melting Pt: 160
Density: 1.145 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00050711
CAS Number: 6865-35-6
EINECS: 229-966-3
UN: 1564
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR