Barium titanate 99

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

93-5640-1000G 93-5640-250G
100202-204EA 145.78 USD
100202-204 100204-372
Barium titanate 99
Barium titanate
Formula: BaO₃Ti
MW: 233.21 g/mol
Melting Pt: 1625
Density: 5.85
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1 III
Merck Index: 12,01029

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR