Barium peroxide ≥84% (99%-Ba)

Supplier: STREM CHEMICALS INC MS
Danger

93-5630-250G 93-5630-1000G
100204-370EA 72.18 USD
100204-370 100199-626
Barium peroxide ≥84% (99%-Ba)
Barium peroxide
Formula: BaO₂
MW: 169.33 g/mol
Melting Pt: 450
Density: 4.96
Storage Temperature: Ambient
MDL Number: MFCD00003454
CAS Number: 1304-29-6
EINECS: 215-128-4
UN: 1449
ADR: 5.1 II
Merck Index: 12,01018

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR