Barium nitrate ≥99.95% (Ba basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5624-50G
93-5624-10G
100208-194EA
142.95
USD
100208-194
100199-140
Barium nitrate ≥99.95% (Ba basis)
Barium nitrate
Formula:
Ba(NO₃)₂ MW: 261.34 g/mol Melting Pt: 592 Density: 3.230 Storage Temperature: Ambient |
MDL Number:
MFCD00003442 CAS Number: 10022-31-8 EINECS: 233-020-5 UN: 1446 ADR: 5.1 II Merck Index: 13,00986 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...