Barium fluoride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-5614-250G 93-5614-1000G
100197-904EA 158.52 USD
100197-904 100211-272
Barium fluoride ≥99%
Barium fluoride
Formula: BaF₂
Boiling Pt: 2137 °C
Melting Pt: 1280 °C
Density: 4.89
Storage Temperature: Ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,01003

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR