Barium fluoride ≥99.99% (Ba basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-5613-50G 93-5613-10G
100205-892EA 462.83 USD
100205-892 100210-484
Barium fluoride ≥99.99% (Ba basis), PURATREM
Barium fluoride
Formula: BaF₂
MW: 175.33 g/mol
Boiling Pt: 2137 °C
Melting Pt: 1280 °C
Density: 4.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,01003

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR