Cesium ≥99.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Prescored ampoule.
93-5550-5G
93-5550-1G
100198-416EA
568.98
USD
100198-416
100197-902
Cesium ≥99.5%
Cesium
Formula:
Cs MW: 132.91 Boiling Pt: 705 Melting Pt: 28.5 Density: 1.892 |
MDL Number:
MFCD00134037 CAS Number: 7440-46-2 EINECS: 231-155-4 UN: 1407 ADR: 4.3 I Merck Index: 12,02051 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...